Tisk…

2-Cyclopropylaniline

Dodavatel: Apollo Scientific
Varování

OR307255-1G OR307255-5G
APOSOR307255-1GEA 5890 CZK
APOSOR307255-1G APOSOR307255-5G
2-Cyclopropylaniline
2-Cyclopropylaniline
Vzorec: C₉H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD00604193
CAS číslo: 3158-73-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR