Tisk…

2-Methyl-2H-indazole-7-carboxylic acid methyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-methyl-2H-indazole-7-carboxylate

OR30725-250MG OR30725-500MG
APOSOR30725-250MGEA 870 CZK
APOSOR30725-250MG APOSOR30725-500MG
2-Methyl-2H-indazole-7-carboxylic acid methyl ester
2-Methyl-2H-indazole-7-carboxylic acid methyl ester
Vzorec: C₁₀H₁₀N₂O₂
MW: 190,2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109408
CAS číslo: 1092351-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR