1-Methyl-1H-indazole-7-carboxylic acid methyl ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 1-methyl-1H-indazole-7-carboxylate
OR30721-1G
OR30721-250MG
OR30721-5G
APOSOR30721-1GEA
7670
CZK
APOSOR30721-1G
APOSOR30721-250MG
APOSOR30721-5G
1-Methyl-1H-indazole-7-carboxylic acid methyl ester
1-Methyl-1H-indazole-7-carboxylic acid methyl ester
Vzorec:
C₁₀H₁₀N₂O₂ MW: 190,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109404 CAS číslo: 1092351-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...