Tisk…

Methyl-1H-indazole-6-carboxylate

Dodavatel: Apollo Scientific
Varování

OR30716-1G OR30716-5G
APOSOR30716-1GEA 622 CZK
APOSOR30716-1G APOSOR30716-5G
Methyl-1H-indazole-6-carboxylate
Methyl-1H-indazole-6-carboxylate
Vzorec: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07371612
CAS číslo: 170487-40-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR