Tisk…

4-Methyl-1-cyclohexanecarbonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Methylcyclohexanecarbonyl chloride

OR307148-1G OR307148-250MG OR307148-500MG
APOSOR307148-1GEA 18620 CZK
APOSOR307148-1G APOSOR307148-250MG APOSOR307148-500MG
4-Methyl-1-cyclohexanecarbonyl chloride
4-Methyl-1-cyclohexanecarbonyl chloride
Vzorec: C₈H₁₃ClO
MW: 160,65 g/mol
Bod varu: 68…72 °C/12mmHg
Hustota: 1,038 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD24230960
CAS číslo: 55930-23-9
UN: 2922
ADR: 8+6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR