Tisk…

(S)-1-(Boc-amino)-2-propanol 98%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl (S)-(2-hydroxypropyl)carbamate

OR307132-100G OR307132-25G OR307132-5G
APOSOR307132-100GEA 10350 CZK
APOSOR307132-100G APOSOR307132-25G APOSOR307132-5G
(S)-1-(Boc-amino)-2-propanol 98%
(S)-1-(Boc-amino)-2-propanol
Vzorec: C₈H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD04974340
CAS číslo: 167938-56-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR