(R)-1-(Boc-amino)-2-propanol 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-tert-Butyl (2-hydroxypropyl)carbamate
OR307131-100G
OR307131-25G
OR307131-5G
APOSOR307131-100GEA
12570
CZK
APOSOR307131-100G
APOSOR307131-25G
APOSOR307131-5G
(R)-1-(Boc-amino)-2-propanol 96%
(R)-1-(Boc-amino)-2-propanol
Vzorec:
C₈H₁₇NO₃ MW: 175,23 g/mol Hustota: 1,02 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD04974338 CAS číslo: 119768-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...