3-Bromo-1H-pyrrolo[2,3-c]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-6-azaindole
OR30682-1G
OR30682-250MG
OR30682-5G
APOSOR30682-1GEA
2030
CZK
APOSOR30682-1G
APOSOR30682-250MG
APOSOR30682-5G
3-Bromo-1H-pyrrolo[2,3-c]pyridine
3-Bromo-1H-pyrrolo[2,3-c]pyridine
Vzorec:
C7H5BrN2 MW: 197,0300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06738910 CAS číslo: 67058-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...