Tisk…

4-(1-Naphthyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(1-Naphthyl)benzeneboronic acid

OR30657-25G OR30657-5G OR30657-100G
APOSOR30657-25GEA 1450 CZK
APOSOR30657-25G APOSOR30657-5G APOSOR30657-100G
4-(1-Naphthyl)phenylboronic acid
4-(1-Naphthyl)phenylboronic acid
Vzorec: C₁₆H₁₃BO₂
MW: 248.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669639
CAS číslo: 870774-25-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR