Tisk…

(5-Methylpyridin-2-yl)methanamine

Dodavatel: Apollo Scientific
Nebezpečí

OR306469-1G OR306469-500MG OR306469-5G
APOSOR306469-1GEA 4900 CZK
APOSOR306469-1G APOSOR306469-500MG APOSOR306469-5G
(5-Methylpyridin-2-yl)methanamine
(5-Methylpyridin-2-yl)methanamine
Vzorec: C₇H₁₀N₂
MW: 122.1677 g/mol
Bod varu: 102…105 °C/12mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD08752541
CAS číslo: 45715-08-0
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR