4-Methoxyphenylboronic acid pinacol ester 97%
Dodavatel: Apollo Scientific
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester
OR30615-5G
OR30615-10G
APOSOR30615-5GEA
1870
CZK
APOSOR30615-5G
APOSOR30615-10G
4-Methoxyphenylboronic acid pinacol ester 97%
4-Methoxyphenylboronic acid pinacol ester
Vzorec:
C₁₃H₁₉BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD05663864 CAS číslo: 171364-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...