3-Bromothieno[2,3-b]pyridine 97%
Dodavatel: Apollo Scientific
OR30580-1G
OR30580-250MG
OR30580-5G
APOSOR30580-1GEA
2240
CZK
APOSOR30580-1G
APOSOR30580-250MG
APOSOR30580-5G
3-Bromothieno[2,3-b]pyridine 97%
3-Bromothieno[2,3-b]pyridine
Vzorec:
C7H4BrNS MW: 214,0824 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11869739 CAS číslo: 28988-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...