Thieno[2,3-b]pyridin-3-ylamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Aminothieno[2,3-b]pyridine
OR30577-1G
OR30577-250MG
OR30577-100MG
APOSOR30577-1GEA
9920
CZK
APOSOR30577-1G
APOSOR30577-250MG
APOSOR30577-100MG
Thieno[2,3-b]pyridin-3-ylamine
Thieno[2,3-b]pyridin-3-ylamine
Vzorec:
C₇H₆N₂S Bod tání: 96…98 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11869737 CAS číslo: 26579-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...