1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR305633-1G
APOSOR305633-1GEA
2200
CZK
APOSOR305633-1G
1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid
1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid
Vzorec:
C₇H₅N₃O₂ MW: 163,1300 g/mol Bod tání: 265…270 °C Storage Temperature: Ambient |
MDL Number:
MFCD11040725 CAS číslo: 116855-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...