Tisk…

Isoquinoline-8-carbaldehyde

Dodavatel: Apollo Scientific
Varování

OR30562-1G OR30562-500MG
APOSOR30562-1GEA 1820 CZK
APOSOR30562-1G APOSOR30562-500MG
Isoquinoline-8-carbaldehyde
Isoquinoline-8-carbaldehyde
Vzorec: C₁₀H₇NO
MW: 157,1687 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06227440
CAS číslo: 787615-01-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR