2-Pyrrolidin-1-ylisonicotinic acid 98%
Dodavatel: Apollo Scientific
OR305419-1G
OR305419-5G
APOSOR305419-1GEA
4860
CZK
APOSOR305419-1G
APOSOR305419-5G
2-Pyrrolidin-1-ylisonicotinic acid 98%
2-Pyrrolidin-1-ylisonicotinic acid
Vzorec:
C₁₀H₁₂N₂O₂ MW: 192.22 g/mol Bod tání: 211…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115212 CAS číslo: 98088-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...