5-Amino-2-chloropyrimidine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR30541-1G
OR30541-5G
APOSOR30541-1GEA
992
CZK
APOSOR30541-1G
APOSOR30541-5G
5-Amino-2-chloropyrimidine 95%
5-Amino-2-chloropyrimidine
Vzorec:
C₄H₄ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD06412560 CAS číslo: 56621-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...