2-Acetamido-6-bromopyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(6-Bromopyridin-2-yl)acetamide
OR305408-1G
OR305408-5G
APOSOR305408-1GEA
2480
CZK
APOSOR305408-1G
APOSOR305408-5G
2-Acetamido-6-bromopyridine
2-Acetamido-6-bromopyridine
Vzorec:
C₇H₇BrN₂O MW: 215,05 g/mol Bod tání: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234277 CAS číslo: 25218-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...