6-Bromo-o-toluic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-6-methylbenzoic acid
OR3054-1G
OR3054-25G
OR3054-5G
APOSOR3054-1GEA
622
CZK
APOSOR3054-1G
APOSOR3054-25G
APOSOR3054-5G
6-Bromo-o-toluic acid 97%
6-Bromo-o-toluic acid
Vzorec:
C₈H₇BrO₂ MW: 215,05 g/mol Bod tání: 107…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD01310788 CAS číslo: 90259-31-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...