4-Bromo-1H-pyrazolo[3,4-c]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR305362-1G
OR305362-250MG
APOSOR305362-1GEA
4140
CZK
APOSOR305362-1G
APOSOR305362-250MG
4-Bromo-1H-pyrazolo[3,4-c]pyridine
4-Bromo-1H-pyrazolo[3,4-c]pyridine
Vzorec:
C₆H₄BrN₃ MW: 198,0201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109831 CAS číslo: 1032943-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...