Tisk…

5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 5-amino-2,3-dihydro-1H-indole-1-carboxylate, tert-Butyl 5-aminoindoline-1-carboxylate

OR305256-1G
APOSOR305256-1GEA 1700 CZK
APOSOR305256-1G
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
Vzorec: C₁₃H₁₈N₂O₂
MW: 234,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059270
CAS číslo: 129487-92-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR