N-Boc-5-hydroxyindole
Dodavatel: Apollo Scientific
Synonyms:
N-Boc-5-Hydroxyindole, tert-Butyl 5-hydroxy-1H-indole-1-carboxylate
OR305227-1G
OR305227-250MG
APOSOR305227-1GEA
2900
CZK
APOSOR305227-1G
APOSOR305227-250MG
N-Boc-5-hydroxyindole
N-Boc-5-hydroxyindole
Vzorec:
C₁₃H₁₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD08272246 CAS číslo: 434958-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...