3-Methylisatoic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
8-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione
OR30520-1G
OR30520-25G
OR30520-5G
APOSOR30520-1GEA
622
CZK
APOSOR30520-1G
APOSOR30520-25G
APOSOR30520-5G
3-Methylisatoic anhydride
3-Methylisatoic anhydride
Vzorec:
C₉H₇NO₃ MW: 177,1568 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426399 CAS číslo: 66176-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...