Imidazo[1,2-a]pyridine-8-carbaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR305169-1G
OR305169-250MG
APOSOR305169-1GEA
3770
CZK
APOSOR305169-1G
APOSOR305169-250MG
Imidazo[1,2-a]pyridine-8-carbaldehyde
Imidazo[1,2-a]pyridine-8-carbaldehyde
Vzorec:
C₈H₆N₂O MW: 146,1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06739248 CAS číslo: 136117-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...