Tisk…

6-Aminoimidazo[1,2-a]pyridine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Imidazo[1,2-a]pyridin-6-amine

OR305164-100MG OR305164-1G OR305164-5G
APOSOR305164-100MGEA 622 CZK
APOSOR305164-100MG APOSOR305164-1G APOSOR305164-5G
6-Aminoimidazo[1,2-a]pyridine 95%
6-Aminoimidazo[1,2-a]pyridine
Vzorec: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD06739232
CAS číslo: 235106-53-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR