1H-Indazole-5-sulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR305122-100MG
OR305122-250MG
OR305122-1G
APOSOR305122-100MGEA
4900
CZK
APOSOR305122-100MG
APOSOR305122-250MG
APOSOR305122-1G
1H-Indazole-5-sulphonyl chloride 97%
1H-Indazole-5-sulphonyl chloride
Vzorec:
C₇H₅ClN₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD11650650 CAS číslo: 599183-35-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...