Tisk…

3-Iodo-6-nitro-1H-indazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Iodo-6-nitroindazole

OR305075-1G OR305075-5G
APOSOR305075-1GEA 827 CZK
APOSOR305075-1G APOSOR305075-5G
3-Iodo-6-nitro-1H-indazole
3-Iodo-6-nitro-1H-indazole
Vzorec: C₇H₄IN₃O₂
MW: 289,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781652
CAS číslo: 70315-70-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR