Hexyl chloroformate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Hexyl carbonochloridate
OR30504-100G
OR30504-25G
APOSOR30504-100GEA
3940
CZK
APOSOR30504-100G
APOSOR30504-25G
Hexyl chloroformate 98%
Hexyl chloroformate
Vzorec:
C₇H₁₃ClO₂ MW: 164.6299 g/mol Bod varu: 60…61 °C/7mmHg Hustota: 1.007 g/cm³ (25 °C) Bod vzplanutí: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000651 CAS číslo: 6092-54-2 EINECS: 228-036-4 UN: 3277 ADR: 6.1+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...