Tisk…

N-Boc-m-phenylenediamine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic acid tert-Butyl ester, N-Boc-1,3-phenylenediamine, 3-(tert-Butoxycarbonylamino)aniline, N-(tert-Butoxycarbonyl)-1,3-phenylenediamine, tert-Butyl (3-aminophenyl)carbamate, 3-(Boc-amino)aniline

OR305002-100G
APOSOR305002-100GEA 4440 CZK
APOSOR305002-100G
N-Boc-m-phenylenediamine 98%
N-Boc-m-phenylenediamine
Vzorec: C₁₁H₁₆N₂O₂
MW: 208,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657915
CAS číslo: 68621-88-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR