5-Chloroisatoic anhydride 97%
Dodavatel: Apollo Scientific
Synonyms:
6-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione
OR30482-100G
APOSOR30482-100GEA
4570
CZK
APOSOR30482-100G
5-Chloroisatoic anhydride 97%
5-Chloroisatoic anhydride
Vzorec:
C₈H₄ClNO₃ MW: 197,58 g/mol Bod tání: 300 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00006701 CAS číslo: 4743-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...