Tisk…

2-Aminoterephthalic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Aminoterephthalic Acid

OR30450-100G OR30450-25G
APOSOR30450-100GEA 8570 CZK
APOSOR30450-100G APOSOR30450-25G
2-Aminoterephthalic acid
2-Aminoterephthalic acid
Vzorec: C₈H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00134536
CAS číslo: 10312-55-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR