4-Aminophthalic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR30446-100G
APOSOR30446-100GEA
2480
CZK
APOSOR30446-100G
4-Aminophthalic acid 96%
4-Aminophthalic acid
Vzorec:
C₈H₇NO₄ MW: 181,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013985 CAS číslo: 5434-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...