Tisk…

Cyclobutyl methyl ketone

Dodavatel: Apollo Scientific

Synonyms: Acetylcyclobutane, Ketone cyclobutyl methyl, 1-Cyclobutylethan-1-one, 1-Cyclobutylethanone, Cyclobutyl Methyl Ketone

OR304351-25G OR304351-5G
APOSOR304351-25GEA 15180 CZK
APOSOR304351-25G APOSOR304351-5G
Cyclobutyl methyl ketone
Cyclobutyl methyl ketone
Vzorec: C₆H₁₀O
MW: 98,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037148
CAS číslo: 3019-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR