L-Homoserine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine
OR304076-25G
OR304076-100G
OR304076-250G
APOSOR304076-25GEA
2360
CZK
APOSOR304076-25G
APOSOR304076-100G
APOSOR304076-250G
L-Homoserine 98%
L-Homoserine
Vzorec:
C₄H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00063090 CAS číslo: 672-15-1 EINECS: 211-590-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...