(S)-(+)-2-Octanol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
D(+)-2-Octanol, (S)-Octan-2-ol, (S)-(+)-Hexyl methyl carbinol, (S)-(+)-2-Hydroxy octane, (S)-(+)-Methyl hexyl carbinol
OR304045-1G
OR304045-5G
APOSOR304045-1GEA
1540
CZK
APOSOR304045-1G
APOSOR304045-5G
(S)-(+)-2-Octanol
(S)-(+)-2-Octanol
Vzorec:
C₈H₁₈O MW: 130,23 g/mol Bod varu: 179…180 °C Bod vzplanutí: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064283 CAS číslo: 6169-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...