Tisk…

2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one

Dodavatel: Apollo Scientific

Synonyms: 2,2,2-Trichloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone

OR303951-1G OR303951-500MG
APOSOR303951-1GEA 7670 CZK
APOSOR303951-1G APOSOR303951-500MG
2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one
2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one
Vzorec: C₉H₅Cl₃N₂O
MW: 263,51 g/mol
Bod tání: 269…279 °C
Storage Temperature: Ambient
MDL Number: MFCD18642801
CAS číslo: 163220-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR