Tisk…

Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate

Dodavatel: Apollo Scientific

OR303948-1G OR303948-500MG
APOSOR303948-1GEA 7670 CZK
APOSOR303948-1G APOSOR303948-500MG
Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate
Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate
Vzorec: C₁₂H₁₃N₃O₄
MW: 263.26 g/mol
Bod tání: 136…137 °C
Storage Temperature: Ambient
MDL Number: MFCD22126111
CAS číslo: 1373348-96-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR