Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 4-(1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)butanoate
OR303932-1G
OR303932-500MG
OR303932-5G
APOSOR303932-1GEA
7670
CZK
APOSOR303932-1G
APOSOR303932-500MG
APOSOR303932-5G
Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0
Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaen-3-yl}butanoate
Vzorec:
C₁₈H₁₇NO₄ MW: 311,34 g/mol Bod tání: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00392161 CAS číslo: 150705-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...