Tisk…

3-Pyridinepropanol

Dodavatel: Apollo Scientific

OR303919-1G OR303919-5G
APOSOR303919-1GEA 0 CZK
APOSOR303919-1G APOSOR303919-5G
3-Pyridinepropanol
3-Pyridinepropanol
Vzorec: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00006414
CAS číslo: 2859-67-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR