Tisk…

5-Chloro-2-(chloromethyl)pyridine

Dodavatel: Apollo Scientific
Nebezpečí

OR303892-1G OR303892-5G OR303892-250MG
APOSOR303892-1GEA 5200 CZK
APOSOR303892-1G APOSOR303892-5G APOSOR303892-250MG
5-Chloro-2-(chloromethyl)pyridine
5-Chloro-2-(chloromethyl)pyridine
Vzorec: C₆H₅Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD10697588
CAS číslo: 10177-24-9
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR