2,3-Dichloroisonicotinonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3-Dichloroisonicotinonitrile
OR303883-500MG
OR303883-1G
APOSOR303883-500MGEA
15940
CZK
APOSOR303883-500MG
APOSOR303883-1G
2,3-Dichloroisonicotinonitrile
2,3-Dichloroisonicotinonitrile
Vzorec:
C₆H₂Cl₂N₂ MW: 173 g/mol Bod tání: 77…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD13193096 CAS číslo: 184416-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...