Tisk…

3-Acetyl-2-pyridine carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Acetyl-2-pyridinecarboxylic acid

OR303851-1G OR303851-500MG
APOSOR303851-1GEA 25460 CZK
APOSOR303851-1G APOSOR303851-500MG
3-Acetyl-2-pyridine carboxylic acid
3-Acetyl-2-pyridine carboxylic acid
Vzorec: C₈H₇NO₃
MW: 165,15 g/mol
Bod tání: 131 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD16140257
CAS číslo: 716362-04-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR