Tisk…

Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate

Dodavatel: Apollo Scientific

OR303807-1G OR303807-500MG
APOSOR303807-1GEA 19110 CZK
APOSOR303807-1G APOSOR303807-500MG
Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate
Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate
Vzorec: C₁₂H₁₂N₂O₂
Bod tání: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD16140350
CAS číslo: 1221791-78-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR