Bicyclo[4.4.0]-2-decanone
Dodavatel: Apollo Scientific
Synonyms:
perhydronaphthalen-2-one
OR30379-1G
OR30379-10G
APOSOR30379-1GEA
2980
CZK
APOSOR30379-1G
APOSOR30379-10G
Bicyclo[4.4.0]-2-decanone
Bicyclo[4.4.0]-2-decanone
Vzorec:
C₁₀H₁₆O MW: 152,24 g/mol Storage Temperature: Ambient |
CAS číslo:
4832-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...