2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Boc-DL-3-(2-Pyridyl)-Ala-OH, 2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
OR303786-1G
OR303786-500MG
APOSOR303786-1GEA
7440
CZK
APOSOR303786-1G
APOSOR303786-500MG
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid
Vzorec:
C₁₃H₁₈N₂O₄ MW: 266.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02094442 CAS číslo: 119434-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...