4-Iodo-2-methoxynicotinaldehyde
Dodavatel: Apollo Scientific
Synonyms:
4-Iodo-2-methoxypyridine-3-carboxaldehyde
OR303775-5G
APOSOR303775-5GEA
2240
CZK
APOSOR303775-5G
4-Iodo-2-methoxynicotinaldehyde
4-Iodo-2-methoxynicotinaldehyde
Vzorec:
C₇H₆INO₂ MW: 263,03 g/mol Bod tání: 96…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD05662396 CAS číslo: 158669-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...