Tisk…

Methyl-2,5-dichloroisonicotinate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2,5-dichloroisonicotinate, Methyl 2,5-dichloropyridine-4-carboxylate

OR303768-1G OR303768-5G
APOSOR303768-1GEA 1580 CZK
APOSOR303768-1G APOSOR303768-5G
Methyl-2,5-dichloroisonicotinate 97%
Methyl-2,5-dichloroisonicotinate
Vzorec: C₇H₅Cl₂NO₂
MW: 206,0300 g/mol
Bod tání: 28…29 °C
Storage Temperature: Ambient
MDL Number: MFCD09951944
CAS číslo: 623585-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR