Tisk…

5-(Cyanomethyl)-2-pyridine carboxylic acid

Dodavatel: Apollo Scientific

OR303739-1G OR303739-500MG
APOSOR303739-1GEA 18550 CZK
APOSOR303739-1G APOSOR303739-500MG
5-(Cyanomethyl)-2-pyridine carboxylic acid
5-(Cyanomethyl)-2-pyridine carboxylic acid
Vzorec: C₈H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16140276
CAS číslo: 1221791-92-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR