Methyl-2,3-dichloroisonicotinate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2,3-dichloroisonicotinate
OR303721-1G
OR303721-5G
APOSOR303721-1GEA
3870
CZK
APOSOR303721-1G
APOSOR303721-5G
Methyl-2,3-dichloroisonicotinate 95%
Methyl-2,3-dichloroisonicotinate
Vzorec:
C₇H₅Cl₂NO₂ MW: 206,0300 g/mol Bod tání: 38…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD09951946 CAS číslo: 603124-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...