Tisk…

tert-Butyl-6-aminopyridin-2-ylcarbamate 95+%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-6-(tert-butoxycarbonylamino)pyridine, 2-Amino-6-(Boc-amino)pyridine, tert-Butyl 6-aminopyridin-2-ylcarbamate

OR303672-1G OR303672-5G OR303672-10G
APOSOR303672-1GEA 6910 CZK
APOSOR303672-1G APOSOR303672-5G APOSOR303672-10G
tert-Butyl-6-aminopyridin-2-ylcarbamate 95+%
tert-Butyl-6-aminopyridin-2-ylcarbamate
Vzorec: C₁₀H₁₅N₃O₂
MW: 209,2500 g/mol
Bod tání: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD09037865
CAS číslo: 322690-31-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR